Assay Details
| Gene Symbol: HUS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079592.1 | NP_001073060.1 | 3 - 4 | 488 | 78 | |
| GenBank mRNA | BC112819.1 | - | 3 - 4 | 488 | 78 | |
More Information
Set Membership: |
|

HUS1
Genomic Map