Assay Details
| Gene Symbol: MED30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079618.2 | NP_001073086.1 | 3 - 4 | 587 | 65 | |
| GenBank mRNA | BC110250.1 | - | 3 - 4 | 588 | 65 | |
More Information
Set Membership: |
|

MED30
Genomic Map