Assay Details
| Gene Symbol: ESX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079624.2 | NP_001073092.1 | 2 - 3 | 542 | 63 | |
| GenBank mRNA | BC109512.1 | - | 2 - 3 | 544 | 63 | |
More Information
Set Membership: |
|

ESX1
Genomic Map