Assay Details
| Gene Symbol: ESX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079624.2 | NP_001073092.1 | 3 - 4 | 589 | 64 | |
| GenBank mRNA | BC109512.1 | - | 3 - 4 | 591 | 64 | |
More Information
Set Membership: |
|

ESX1
Genomic Map