Assay Details
| Gene Symbol: TRIM59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080310.2 | NP_001073779.1 | 2 - 3 | 137 | 130 | |
| GenBank mRNA | BC112616.1 | - | 2 - 3 | 139 | 130 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC112616.1, NM_001080310.2 transcripts.
Set Membership: |
|

TRIM59
Genomic Map