Assay Details
| Gene Symbol: MED29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080316.2 | NP_001073785.1 | 1 - 2 | 216 | 72 | |
| GenBank mRNA | BC126813.1 | - | 1 - 2 | 217 | 72 | |
More Information
Set Membership: |
|

MED29
Genomic Map