Assay Details
| Gene Symbol: GNG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174073.2 | NP_776498.1 | 1 - 2 | 104 | 120 | |
| GenBank mRNA | BC109874.1 | - | 1 - 2 | 104 | 120 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC109874.1, NM_174073.2 transcripts.
Set Membership: |
|

GNG3
Genomic Map