Assay Details
| Gene Symbol: PLAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174146.3 | NP_776571.2 | 12 - 13 | 1468 | 57 | |
| GenBank mRNA | BC105348.1 | - | 12 - 13 | 1468 | 57 | |
| X85800.1 | - | 1382 | 57 | |||
More Information
Set Membership: |
|

PLAT
Genomic Map