Assay Details
| Gene Symbol: EIF4E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174310.3 | NP_776735.2 | 3 - 4 | 233 | 107 | |
| GenBank mRNA | AF257235.1 | - | 3 - 4 | 232 | 107 | |
| BC111272.1 | - | 3 - 4 | 233 | 107 | ||
More Information
Set Membership: |
|

EIF4E
Genomic Map