Assay Details
| Gene Symbol: GNA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174322.4 | NP_776747.1 | 3 - 4 | 710 | 73 | |
| GenBank mRNA | BC111118.1 | - | 3 - 4 | 711 | 73 | |
| D90336.1 | - | 3 - 4 | 705 | 73 | ||
More Information
Set Membership: |
|

GNA11
Genomic Map