Assay Details
| Gene Symbol: PRP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212448.2 | NP_997613.2 | 5 - 6 | 987 | 117 | |
| GenBank mRNA | AB239755.2 | - | 4 - 5 | 561 | 117 | |
| BC109590.1 | - | 5 - 6 | 987 | 117 | ||
More Information
Set Membership: |
|

PRP4
Genomic Map