Assay Details
| Gene Symbol: BOLA-DMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012674.2 | NP_001012692.2 | 1 - 2 | 93 | 96 | |
| GenBank mRNA | BC114012.1 | - | 1 - 2 | 93 | 96 | |
| D76416.1 | - | 1 - 2 | 92 | 96 | ||
More Information
Set Membership: |
|

BOLA-DMA
Genomic Map