Assay Details
| Gene Symbol: TBC1D7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015643.2 | NP_001015643.1 | 3 - 4 | 346 | 69 | |
| GenBank mRNA | BC118160.1 | - | 3 - 4 | 346 | 69 | |
| BT020996.1 | - | 3 - 4 | 432 | 69 | ||
More Information
Set Membership: |
|

TBC1D7
Genomic Map