Assay Details
| Gene Symbol: FAM118B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015677.2 | NP_001015677.1 | 4 - 5 | 631 | 68 | |
| GenBank mRNA | BC118445.1 | - | 4 - 5 | 631 | 68 | |
| BT021043.1 | - | 4 - 5 | 609 | 68 | ||
More Information
Set Membership: |
|

FAM118B
Genomic Map