Assay Details
| Gene Symbol: TANGO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035328.2 | NP_001030405.2 | 3 - 4 | 306 | 63 | |
| GenBank mRNA | BC113297.1 | - | 3 - 4 | 306 | 63 | |
| BT021651.1 | - | 2 - 3 | 572 | 63 | ||
More Information
Set Membership: |
|

TANGO2
Genomic Map