Assay Details
| Gene Symbol: BOLA-DMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040481.3 | NP_001035571.1 | 1 - 2 | 271 | 106 | |
| GenBank mRNA | BC118361.1 | - | 1 - 2 | 272 | 106 | |
| BT021480.1 | - | 1 - 2 | 219 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118361.1, NM_001040481.3 transcripts.
Set Membership: |
|

BOLA-DMB
Genomic Map