Assay Details
| Gene Symbol: BOLA-DMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040481.3 | NP_001035571.1 | 2 - 3 | 551 | 81 | |
| GenBank mRNA | BC118361.1 | - | 2 - 3 | 552 | 81 | |
| BT021480.1 | - | 2 - 3 | 499 | 81 | ||
| D76417.1 | - | 2 - 3 | 335 | 81 | ||
More Information
Set Membership: |
|

BOLA-DMB
Genomic Map