Assay Details
| Gene Symbol: BOLA-DMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040481.3 | NP_001035571.1 | 5 - 6 | 991 | 79 | |
| GenBank mRNA | BC118361.1 | - | 5 - 6 | 992 | 79 | |
| BT021480.1 | - | 5 - 6 | 939 | 79 | ||
More Information
Set Membership: |
|

BOLA-DMB
Genomic Map