Assay Details
| Gene Symbol: DNAJA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040491.1 | NP_001035581.1 | 7 - 8 | 972 | 76 | |
| GenBank mRNA | BC113244.1 | - | 7 - 8 | 972 | 76 | |
More Information
Set Membership: |
|

DNAJA2
Genomic Map