Assay Details
| Gene Symbol: CS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044721.1 | NP_001038186.1 | 5 - 6 | 509 | 67 | |
| GenBank mRNA | AF461103.1 | - | 2 - 3 | 185 | 67 | |
| BC114138.1 | - | 5 - 6 | 509 | 67 | ||
More Information
Set Membership: |
|

CS
Genomic Map