Assay Details
| Gene Symbol: CS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044721.1 | NP_001038186.1 | 9 - 10 | 1131 | 66 | |
| GenBank mRNA | BC114138.1 | - | 9 - 10 | 1131 | 66 | |
| DQ403140.1 | - | 3 - 4 | 351 | 66 | ||
More Information
Set Membership: |
|

CS
Genomic Map