Assay Details
| Gene Symbol: CCNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045872.1 | NP_001039337.1 | 5 - 6 | 757 | 86 | |
| GenBank mRNA | BC116011.1 | - | 5 - 6 | 757 | 86 | |
| L26548.1 | - | 5 - 6 | 806 | 86 | ||
More Information
Set Membership: |
|

CCNB1
Genomic Map