Assay Details
| Gene Symbol: TRNT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045876.2 | NP_001039341.1 | 2 - 3 | 228 | 77 | |
| GenBank mRNA | BC114831.1 | - | 2 - 3 | 229 | 77 | |
More Information
Set Membership: |
|

TRNT1
Genomic Map