Assay Details
| Gene Symbol: PLPP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045890.1 | NP_001039355.1 | 3 - 4 | 540 | 94 | |
| GenBank mRNA | BC113292.1 | - | 3 - 4 | 540 | 94 | |
More Information
Set Membership: |
|

PLPP2
Genomic Map