Assay Details
| Gene Symbol: ZNHIT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045895.1 | NP_001039360.1 | 2 - 3 | 166 | 65 | |
| GenBank mRNA | BC112640.1 | - | 2 - 3 | 166 | 65 | |
More Information
Set Membership: |
|

ZNHIT3
Genomic Map