Assay Details
| Gene Symbol: KDSR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045919.1 | NP_001039384.1 | 6 - 7 | 653 | 65 | |
| GenBank mRNA | BC112615.1 | - | 6 - 7 | 653 | 65 | |
More Information
Set Membership: |
|

KDSR
Genomic Map