Assay Details
| Gene Symbol: RSAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045941.1 | NP_001039406.1 | 2 - 3 | 566 | 57 | |
| GenBank mRNA | BC105455.1 | - | 2 - 3 | 566 | 57 | |
More Information
Set Membership: |
|

RSAD2
Genomic Map