Assay Details
| Gene Symbol: RND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046016.1 | NP_001039481.1 | 1 - 2 | 191 | 66 | |
| GenBank mRNA | BC113283.1 | - | 1 - 2 | 191 | 66 | |
More Information
Set Membership: |
|

RND1
Genomic Map