Assay Details
| Gene Symbol: NAPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046022.2 | NP_001039487.1 | 2 - 3 | 198 | 62 | |
| GenBank mRNA | BC114119.1 | - | 2 - 3 | 199 | 62 | |
More Information
Set Membership: |
|

NAPG
Genomic Map