Assay Details
| Gene Symbol: PTDSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046040.1 | NP_001039505.1 | 9 - 10 | 1084 | 60 | |
| GenBank mRNA | BC112833.1 | - | 9 - 10 | 1084 | 60 | |
More Information
Set Membership: |
|

PTDSS1
Genomic Map