Assay Details
| Gene Symbol: RTCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046055.1 | NP_001039520.1 | 6 - 7 | 793 | 61 | |
| GenBank mRNA | BC113251.1 | - | 6 - 7 | 793 | 61 | |
More Information
Set Membership: |
|

RTCA
Genomic Map