Assay Details
| Gene Symbol: ADH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046057.2 | NP_001039522.1 | 1 - 2 | 45 | 72 | |
| GenBank mRNA | BC112630.1 | - | 1 - 2 | 46 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC112630.1, NM_001046057.2 transcripts.
Set Membership: |
|

ADH6
Genomic Map