Assay Details
| Gene Symbol: ADH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046057.2 | NP_001039522.1 | 5 - 6 | 593 | 66 | |
| GenBank mRNA | BC112630.1 | - | 5 - 6 | 594 | 66 | |
More Information
Set Membership: |
|

ADH6
Genomic Map