Assay Details
| Gene Symbol: ADH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046057.2 | NP_001039522.1 | 8 - 9 | 1129 | 126 | |
| GenBank mRNA | BC112630.1 | - | 8 - 9 | 1130 | 126 | |
More Information
Set Membership: |
|

ADH6
Genomic Map