Assay Details
| Gene Symbol: LCAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046069.2 | NP_001039534.1 | 3 - 4 | 442 | 72 | |
| GenBank mRNA | BC112484.1 | - | 3 - 4 | 443 | 72 | |
More Information
Set Membership: |
|

LCAT
Genomic Map