Assay Details
| Gene Symbol: ATP6V0D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046101.1 | NP_001039566.1 | 6 - 7 | 950 | 62 | |
| GenBank mRNA | BC105425.1 | - | 6 - 7 | 950 | 62 | |
More Information
Set Membership: |
|

ATP6V0D2
Genomic Map