Assay Details
| Gene Symbol: MALL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046115.2 | NP_001039580.1 | 2 - 3 | 298 | 83 | |
| GenBank mRNA | BC105332.1 | - | 2 - 3 | 299 | 83 | |
More Information
Set Membership: |
|

MALL
Genomic Map