Assay Details
| Gene Symbol: MALL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046115.2 | NP_001039580.1 | 3 - 4 | 421 | 76 | |
| GenBank mRNA | BC105332.1 | - | 3 - 4 | 422 | 76 | |
More Information
Set Membership: |
|

MALL
Genomic Map