Assay Details
| Gene Symbol: MRPS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046123.2 | NP_001039588.1 | 6 - 7 | 685 | 81 | |
| GenBank mRNA | BC112674.1 | - | 6 - 7 | 686 | 81 | |
More Information
Set Membership: |
|

MRPS5
Genomic Map