Assay Details
| Gene Symbol: SH3GL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046130.2 | NP_001039595.1 | 3 - 4 | 384 | 67 | |
| GenBank mRNA | BC105444.1 | - | 3 - 4 | 385 | 67 | |
| BT025352.1 | - | 3 - 4 | 267 | 67 | ||
More Information
Set Membership: |
|

SH3GL1
Genomic Map