Assay Details
| Gene Symbol: PHF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046144.1 | NP_001039609.1 | 6 - 7 | 777 | 88 | |
| GenBank mRNA | BC113245.1 | - | 6 - 7 | 777 | 88 | |
More Information
Set Membership: |
|

PHF11
Genomic Map