Assay Details
| Gene Symbol: PMVK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046151.2 | NP_001039616.1 | 1 - 2 | 113 | 54 | |
| GenBank mRNA | BC112509.1 | - | 1 - 2 | 118 | 54 | |
More Information
Set Membership: |
|

PMVK
Genomic Map