Assay Details
| Gene Symbol: C14H8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC112675.1 | - | 3 - 4 | 222 | 80 | |
| EH210109.1 | - | 3 - 4 | 239 | 80 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C14H8orf33| Gene Symbol: C14H8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC112675.1 | - | 3 - 4 | 222 | 80 | |
| EH210109.1 | - | 3 - 4 | 239 | 80 | ||
Set Membership: |
|