Assay Details
| Gene Symbol: C14H8orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046153.3 | NP_001039618.2 | 5 - 6 | 469 | 124 | |
| GenBank mRNA | BC112675.1 | - | 5 - 6 | 453 | 124 | |
| CK771102.1 | - | 1 - 2 | 762 | 124 | ||
| EH210109.1 | - | 5 - 6 | 470 | 124 | ||
More Information
Set Membership: |
|

C14H8orf33
Genomic Map