Assay Details
| Gene Symbol: ARG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046154.1 | NP_001039619.1 | 2 - 3 | 199 | 68 | |
| GenBank mRNA | BC105497.1 | - | 2 - 3 | 199 | 68 | |
More Information
Set Membership: |
|

ARG1
Genomic Map