Assay Details
| Gene Symbol: EMG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046197.2 | NP_001039662.2 | 1 - 2 | 301 | 108 | |
| GenBank mRNA | BC113268.1 | - | 1 - 2 | 276 | 108 | |
| DV843519.1 | - | 1 - 2 | 302 | 108 | ||
| EV745816.1 | - | 5 - 6 | 741 | 108 | ||
More Information
Set Membership: |
|

EMG1
Genomic Map