Assay Details
| Gene Symbol: RAB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046209.1 | NP_001039674.1 | 1 - 2 | 356 | 63 | |
| GenBank mRNA | BC114066.1 | - | 1 - 2 | 356 | 63 | |
More Information
Set Membership: |
|

RAB26
Genomic Map