Assay Details
| Gene Symbol: LYNX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046221.2 | NP_001039686.1 | 1 - 2 | 74 | 74 | |
| GenBank mRNA | BC114776.1 | - | 1 - 2 | 76 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC114776.1, NM_001046221.2 transcripts.
Set Membership: |
|

LYNX1
Genomic Map