Assay Details
| Gene Symbol: LYNX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046221.2 | NP_001039686.1 | 3 - 4 | 377 | 69 | |
| GenBank mRNA | BC114776.1 | - | 3 - 4 | 379 | 69 | |
More Information
Set Membership: |
|

LYNX1
Genomic Map