Assay Details
| Gene Symbol: GPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC105496.1 | - | 7 - 8 | 1436 | 69 | |
| BC153219.1 | - | 7 - 8 | 1465 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPC1| Gene Symbol: GPC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC105496.1 | - | 7 - 8 | 1436 | 69 | |
| BC153219.1 | - | 7 - 8 | 1465 | 69 | ||
Set Membership: |
|